$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 5,6-Dehydro-17Beta-dutasteride
TRC 5,6-Dehydro-17Beta-dutasteride
TRC 5,6-Dehydro-17Beta-dutasteride
Catalog # 77981-305
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 5,6-Dehydro-17Beta-dutasteride
Catalog # 77981-305
Supplier:
LGC Standards
Supplier Number:
TRC-D229560-5MG
New Product
CAS Number:
Specifications
CAS No.:
1430804-85-5
Description:
5,6-Dehydro-17Beta-dutasteride
Size:
5 mg
Specifications
About this item
Product Details
Product Details