Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
TRC N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
TRC N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
Catalog # 77984-275
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
Catalog # 77984-275
Supplier:
LGC Standards
Supplier Number:
TRC-E898280-10MG
New Product
CAS Number:
Specifications
CAS No.:
1217262-11-7
Description:
N-(Ethylamino) Fluvoxamine (up to 15% Z isomer)
Size:
10 mg
Specifications
About this item
Product Details
Product Details