$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Dimethyldimyristylammonium Chloride
TRC Dimethyldimyristylammonium Chloride
77983-096
:
LGC Standards
New Product
:
TRC Dimethyldimyristylammonium Chloride
77983-096
:
LGC Standards
:
TRC-D472560-250MG
New Product
:
CAS No.:
10108-91-5
Description:
Dimethyldimyristylammonium Chloride
Size:
250 mg