TRC 6Alpha-Bromo Beclomethasone Dipropionate
New Product
- CAS No.:887130-69-0
Frequently Bought Together
CrossLab LC/MSD Supplies for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Needle and Needle Seats for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
: Thermo Scientific
CrossLab Sampler and Maintenance Kits for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Torches for 5100/5110 and 5800/5900 ICP-OES, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
O-Rings and Fittings for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)