Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 1-Acetyl-4-(4-hydroxyphenyl)piperazine
TRC 1-Acetyl-4-(4-hydroxyphenyl)piperazine
TRC 1-Acetyl-4-(4-hydroxyphenyl)piperazine
77976-596
:
LGC Standards
New Product
:
TRC 1-Acetyl-4-(4-hydroxyphenyl)piperazine
77976-596
:
LGC Standards
:
TRC-A179045-1G
New Product
:
CAS No.:
67914-60-7
Description:
1-Acetyl-4-(4-hydroxyphenyl)piperazine
Size:
1 g