$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (S)-Remoxipride Hydrochloride
TRC (S)-Remoxipride Hydrochloride
TRC (S)-Remoxipride Hydrochloride
Catalog # 77992-667
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC (S)-Remoxipride Hydrochloride
Catalog # 77992-667
Supplier:
LGC Standards
Supplier Number:
TRC-R143700-10MG
New Product
CAS Number:
Specifications
CAS No.:
73220-03-8
Description:
(S)-Remoxipride Hydrochloride
Size:
10 mg
Specifications
About this item
Product Details
Product Details