$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Isophorone Diamine-d5 (Major) Dihydrochloride Salt (cis/trans Mixture)
TRC Isophorone Diamine-d5 (Major) Dihydrochloride Salt (cis/trans Mixture)
77987-398
:
LGC Standards
New Product
:
TRC Isophorone Diamine-d5 (Major) Dihydrochloride Salt (cis/trans Mixture)
77987-398
:
LGC Standards
:
TRC-I821937-1MG
New Product
:
CAS No.:
1794752-32-1
Description:
Isophorone Diamine-d5 (Major) Dihydrochloride Salt (cis/trans Mixture)
Size:
1 mg