$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Sodium O,O-Dimethyl Thiophosphate (Technical grade)
TRC Sodium O,O-Dimethyl Thiophosphate (Technical grade)
77993-246
:
LGC Standards
New Product
:
TRC Sodium O,O-Dimethyl Thiophosphate (Technical grade)
77993-246
:
LGC Standards
:
TRC-S624240-250MG
New Product
:
CAS No.:
23754-87-2
Description:
Sodium O,O-Dimethyl Thiophosphate (Technical grade)
Size:
250 mg