$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 5-Chloro-2-methyl-3-isothiazolone (~90%)
TRC 5-Chloro-2-methyl-3-isothiazolone (~90%)
:
LGC Standards
New Product
TRC 5-Chloro-2-methyl-3-isothiazolone (~90%)
:
LGC Standards
New Product
CAS No.:
26172-55-4