Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Hydrazine HR Visual Test Kit 0 to 500 ppm
CHEMets® Hydrazine HR Visual Test Kit 0 to 500 ppm
Catalog # 77674-782
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Hydrazine HR Visual Test Kit 0 to 500 ppm
Catalog # 77674-782
Supplier:  CHEMetrics
Supplier Number:  K-5020C
New Product
CAS Number:  

Specifications

  • Chemical method:
    PDMAB
  • Application:
    Boiler feed water;Process water

Specifications

About this item

Delivery: The K-5020C hydrazine HR CHEMets® test kit (0 to 500 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): R to 5005 CHEMets®refill, C to 5015C comparator, 25 ml sample cup, A to 0171 Pipettor Tips Pack, A to 0191 MiniPet®, and instructions.