$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Tris(2-ethylhexyl) Phosphate
TRC Tris(2-ethylhexyl) Phosphate
Catalog # 77994-897
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Tris(2-ethylhexyl) Phosphate
Catalog # 77994-897
Supplier:
LGC Standards
Supplier Number:
TRC-T884650-10G
New Product
CAS Number:
Specifications
CAS No.:
78-42-2
Description:
Tris(2-ethylhexyl) Phosphate
Size:
10 g
Specifications
About this item
Product Details
Product Details