$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride
TRC L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride
77988-656
:
LGC Standards
New Product
:
TRC L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride
77988-656
:
LGC Standards
:
TRC-M260472-2.5MG
New Product
:
CAS No.:
362049-54-5
Description:
L-Methionine-S-methyl Sulfonium Chloride-d6 Hydrochloride
Size:
2.5 mg