TRC Nafion
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:31175-20-9
Specifications
Frequently Bought Together
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
LC Columns for PFC Analysis, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
AVS 6/7 Supplies for ICP-OES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Integrator Supplies and Cables, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
CrossLab Quick Connect and Quick Turn Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Pump Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Fittings for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)