molybdate Instrumental Test 0 to 25 ppm
Catalog # 77674-820
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Catechol
- Application:Industrial water treatment;Power generation;Cooling water systems
Specifications
About this item
- Range: 0 to 25.0 ppm as Mo Method: Catechol
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics direct-readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.