$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Elvitegravir-d6 (Major)
TRC Elvitegravir-d6 (Major)
77983-818
:
LGC Standards
New Product
:
TRC Elvitegravir-d6 (Major)
77983-818
:
LGC Standards
:
TRC-E509002-1MG
New Product
:
CAS No.:
1131640-69-1
Description:
Elvitegravir-d6 (Major)
Size:
1 mg