ContactUsLinkComponent
CHEMets® Nitrite HR Visual Test Kit 0 to 250 ppm
CHEMets® Nitrite HR Visual Test Kit 0 to 250 ppm
  77674-838
New Product
 :  
undefined
CHEMets® Nitrite HR Visual Test Kit 0 to 250 ppm
  77674-838
 :  K-7020B
New Product
 :  
  • Chemical method:
    Azo dye formation
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water
  • Method: Azo dye formation Range: 0 to 250 ppm MDL: 20 ppm
 : The K-7020B Nitrite HR CHEMets® test kit (0 to 250 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets®refill, flat comparator, 25 ml sample cup, pipettor tips, 200 µl fixed volume pipet, and instructions.