Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed 400 mg/dl Aq. Ethanol Standard Solution, 400 mg/dl in Water
Lipomed 400 mg/dl Aq. Ethanol Standard Solution, 400 mg/dl in Water
Catalog # 77976-164
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Lipomed 400 mg/dl Aq. Ethanol Standard Solution, 400 mg/dl in Water
Catalog # 77976-164
Supplier:  LGC Standards
Supplier Number:  LPM-ETH-400-1ML-5AMP
New Product
CAS Number:  

Specifications

  • Matrix:
    Water
  • CAS No.:
    64-17-5
  • Description:
    400 mg/dL Aq. Ethanol Standard Solution, 400 mg/dL in Water
  • Size:
    5×1 ml

Specifications

About this item