Lipomed d,l-EDDP-D3.perchlorate, 1 mg/ml in Methanol (as pyrrolinium)
Catalog # 77976-149
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:136765-23-6
- Description:d,l-EDDP-D3.perchlorate, 1 mg/ml in Methanol (as pyrrolinium)
- Size:1 ml