Paragon Scientific Type CL140 (Cl14), Cold Crank Simulator Viscosity
Catalog # 77975-527
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Type CL140 (Cl14), cold crank simulator viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component