Order Entry
Puerto Rico
ContactUsLinkComponent
TRC 3’-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone(Domperidone Impurity E) (>90%)
TRC 3’-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone(Domperidone Impurity E) (>90%)
Catalog # 77982-773
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC 3’-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone(Domperidone Impurity E) (>90%)
Catalog # 77982-773
Supplier:  LGC Standards
Supplier Number:  TRC-D449465-10MG
New Product
CAS Number:  

Specifications

  • CAS No.:
    1346602-50-3
  • Description:
    3’-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone(Domperidone Impurity E) (>90%)
  • Size:
    10 mg

Specifications

About this item