Lipomed Codeine.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-116
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:1422-07-7
- Description:Codeine.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml