Order Entry
Puerto Rico
ContactUsLinkComponent
 
Paragon Scientific Small Sample Viscosity (13014 cSt @ 40 °C), Certified @ 40 and 100 °C
Catalog # 77975-780
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Paragon Scientific Small Sample Viscosity (13014 cSt @ 40 °C), Certified @ 40 and 100 °C
Catalog # 77975-780
Supplier:  LGC Standards
Supplier Number:  ALK-HVS15
New Product
CAS Number:  

Specifications

  • Description:
    Small sample viscosity (13014 cSt @ 40 °C), certified @ 40 and 100 °C
  • Size:
    60 ml
  • Shipping Conditions:
    RT
  • Storage Temperature:
    RT
  • Certification:
    ISO 17034

Specifications

About this item

  • Standard type: Single component