CHEMets® molybdate Visual Test Kit 2 to 24 ppm
Catalog # 77674-818
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Catechol
- Application:Industrial water treatment;Power generation;Cooling water systems
Specifications
About this item
- Range: 2 to 24 ppm as Mo MDL: 2 ppm Method: Catechol
Delivery: Kit K-6702 comes in a plastic case and contains everything needed to perform 30 tests:refill, comparator, 25 ml sample cup, and instructions.