$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (-)-Strigolactone GR24
TRC (-)-Strigolactone GR24
TRC (-)-Strigolactone GR24
Catalog # 77993-488
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC (-)-Strigolactone GR24
Catalog # 77993-488
Supplier:
LGC Standards
Supplier Number:
TRC-S687610-1MG
New Product
CAS Number:
Specifications
CAS No.:
188062-52-4
Description:
(-)-Strigolactone GR24
Size:
1 mg
Specifications
About this item
Product Details
Product Details