Order Entry
Puerto Rico
ContactUsLinkComponent
DEHA Instrumental Test 0 to 2 ppm
DEHA Instrumental Test 0 to 2 ppm
Catalog # 77674-774
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
DEHA Instrumental Test 0 to 2 ppm
Catalog # 77674-774
Supplier:  CHEMetrics
Supplier Number:  K-3903
New Product
CAS Number:  

Specifications

  • Chemical method:
    PDTS
  • Application:
    Boiler feed water and condensate

Specifications

About this item

  • Range: 0 to 2.00 ppm Method: PDTS
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, activator solution, 25 ml sample cup, ampoule blank, and instructions. This kit requires the use of a CHEMetrics Direct to readout photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately.