Order Entry
Puerto Rico
ContactUsLinkComponent
Paragon Scientific Sulfur @ 0.01% (100 ppm) in odorless kerosene
Paragon Scientific Sulfur @ 0.01% (100 ppm) in odorless kerosene
Catalog # 77975-831
Supplier:  LGC Standards
New Product
CAS Number:  
Paragon Scientific Sulfur @ 0.01% (100 ppm) in odorless kerosene
Catalog # 77975-831
Supplier:  LGC Standards
Supplier Number:  ALK-KRSU-05
New Product
CAS Number:  

Specifications

  • Matrix:
    Odourless Kerosene
  • CAS No.:
    7704-34-9
  • Description:
    Sulfur @ 0.01% (100 ppm) in odorless kerosene
  • Size:
    100 ml
  • Certification:
    ISO 17034
  • Storage Conditions:
    RT
  • Shipping Conditions:
    RT
  • Molecular Weight:
    32.065

Specifications

About this item