TRC Fluticasone Furoate
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:397864-44-7
Specifications
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
CrossLab Dissolution Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Sample Trays for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Capillaries for Waters LC Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Stay Safe Caps for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
LC Columns for PFC Analysis, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Plugs and Caps, IDEX Health and Science
Supplier: Upchurch Scientific
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
ZORBAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Heat Exchanger for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Sealing Mats for InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)