$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (R,R)-Labetalol-d3 Hydrochloride
TRC (R,R)-Labetalol-d3 Hydrochloride
Catalog # 77987-784
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC (R,R)-Labetalol-d3 Hydrochloride
Catalog # 77987-784
Supplier:
LGC Standards
Supplier Number:
TRC-L096502-1MG
New Product
CAS Number:
Specifications
CAS No.:
0
Description:
(R,R)-Labetalol-d3 Hydrochloride
Size:
1 mg
Specifications
About this item
Product Details
Product Details