Order Entry
Puerto Rico
ContactUsLinkComponent
 
Phosphate Comparator Round 0 to 1 ppm
Catalog # 77674-674
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Phosphate Comparator Round 0 to 1 ppm
Catalog # 77674-674
Supplier:  CHEMetrics
Supplier Number:  C-8501
New Product
CAS Number:  

Specifications

  • Chemical method:
    Stannous Chloride

Specifications

About this item

C-8501 Low Range Phosphate, ortho Comparator is a component of K-8510 Phosphate, ortho CHEMets Visual Test Kit and is designed to be compared against the color formed in R-8510 Phosphate, ortho CHEMets Refill. The comparator's range is applicable to many environmental and water treatment applications. Color standards are blue in color and cover a range of 0 to 1.0 ppm PO₄.