Order Entry
Puerto Rico
ContactUsLinkComponent
Carbon Dioxide Titrate Test 250 to 2500 ppm
Carbon Dioxide Titrate Test 250 to 2500 ppm
Catalog # 77674-742
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Carbon Dioxide Titrate Test 250 to 2500 ppm
Catalog # 77674-742
Supplier:  CHEMetrics
Supplier Number:  K-1925
New Product
CAS Number:  

Specifications

  • Chemical method:
    Caustic titrant with pH indicator
  • Application:
    Surface and ground water;Municipal water treatment systems

Specifications

About this item

Features of titrets technology: Dependable, sealed single-use titrants Fast, easy procedure no need to count drops No burettes or other equipment necessary Perform a titration ANYWHERE

  • Range: 250 to 2500 ppm MDL: 250 ppm Method: Caustic titrant with pH indicator Increments: 250, 275, 300, 325, 350, 375, 400, 450, 500, 625, 750, 875, 1000, 1250, 1750, 2500 ppm
Delivery: Kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules with valve assemblies, activator solution, titrettor, 25 ml sample cup, and instructions.