Order Entry
Puerto Rico
ContactUsLinkComponent
Foalwatch (Calcium) Test Kit
Foalwatch (Calcium) Test Kit
Catalog # 77674-736
Supplier:  CHEMetrics
New Product
CAS Number:  
Foalwatch (Calcium) Test Kit
Catalog # 77674-736
Supplier:  CHEMetrics
Supplier Number:  K-1700
New Product
CAS Number:  

Specifications

  • Chemical method:
    EGTA (calcium)
  • Application:
    Drinking water;Highly purified waste water;Sea water

Specifications

About this item

The FoalWatch system is based on proven chemistry, so you can rest easier at night. The test measures the concentration of calcium in the, colostrum. The calcium level rises sharply before birth. Simply begin testing once or twice a day about 2 weeks before the mareís expected foaling date. When the calcium concentration consistently exceeds 200 parts per million (ppm), birth is imminent.FoalWatch Kit contains everything you need for testing.

Safe and easy to use. Use of FoalWatch kits require no specialized training. Allows safe and comfortable movement of mare prior to foaling. Contains everything that is needed for testing FoalWatch kits comes complete with 20 Test Ampoules, test tube for collecting mareís milk, bottle of distilled water, syringe, sample cup, and complete instructions.

Delivery: Kit comes complete with: 20 test ampoules, test tube for collecting mare's milk, bottle of distilled water, syringe, sample cup and complete instructions