$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC o-Flutamide
TRC o-Flutamide
77984-995
:
LGC Standards
New Product
:
TRC o-Flutamide
77984-995
:
LGC Standards
:
TRC-F598855-10MG
New Product
:
CAS No.:
151262-93-0
Description:
o-Flutamide
Size:
10 mg