$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Norfluoxetine-d5 Hydrochloride (Phenyl-d5)
TRC Norfluoxetine-d5 Hydrochloride (Phenyl-d5)
77990-579
:
LGC Standards
New Product
:
TRC Norfluoxetine-d5 Hydrochloride (Phenyl-d5)
77990-579
:
LGC Standards
:
TRC-N681103-1MG
New Product
:
CAS No.:
1188265-34-0
Description:
Norfluoxetine-d5 Hydrochloride (Phenyl-d5)
Size:
1 mg