$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Phospholan-methyl
TRC Phospholan-methyl
77991-725
:
LGC Standards
New Product
:
TRC Phospholan-methyl
77991-725
:
LGC Standards
:
TRC-P358350-50MG
New Product
:
CAS No.:
5120-23-0
Description:
Phospholan-methyl
Size:
50 mg