$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Zonisamide-13C2-15N
TRC Zonisamide-13C2-15N
77995-514
:
LGC Standards
New Product
:
TRC Zonisamide-13C2-15N
77995-514
:
LGC Standards
:
TRC-Z700003-1MG
New Product
:
CAS No.:
1188265-58-8
Description:
Zonisamide-13C2-15N
Size:
1 mg