ContactUsLinkComponent
Ammonia Accessory Solutions Pack
Ammonia Accessory Solutions Pack
Catalog # 77674-532
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Ammonia Accessory Solutions Pack
Catalog # 77674-532
Supplier:  CHEMetrics
Supplier Number:  A-1410
New Product
CAS Number:  
Specifications
  • Chemical method:
    Hydroxybenzyl Alcohol (HBA)
Specifications
About this item
A-1410 is designed to be a refill of the accessory solutions found in the K-1420 Ammonia CHEMets®; Test Kit (HBA). It contains: Two 10 ml bottles of A-1404 Stabilizer Solution, two 10 ml bottles of A-1405 Catalyzer Solution, and two 10 ml bottles of A-1406 Activator Solution. This is enough of each solution to perform approximately 80 tests.