$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Phentolamine Hydrochloride
TRC Phentolamine Hydrochloride
77991-553
:
LGC Standards
New Product
:
TRC Phentolamine Hydrochloride
77991-553
:
LGC Standards
:
TRC-P302550-100MG
New Product
:
CAS No.:
73-05-2
Description:
Phentolamine Hydrochloride
Size:
100 mg