$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Lipomed Buprenorphine.HCl, 1 mg/ml in Methanol (as free base)
Lipomed Buprenorphine.HCl, 1 mg/ml in Methanol (as free base)
77976-103
:
LGC Standards
New Product
:
Lipomed Buprenorphine.HCl, 1 mg/ml in Methanol (as free base)
77976-103
:
LGC Standards
:
LPM-BUP-399-HC-1LM
New Product
:
Matrix:
Methanol
CAS No.:
53152-21-9
Description:
Buprenorphine.HCl, 1 mg/ml in Methanol (as free base)
Size:
1 ml