TRC Atipamezole Hydrochloride
New Product
- CAS No.:104075-48-1
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
CrossLab Macrocells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Syringe Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
: Upchurch Scientific
CrossLab Dissolution Full Flow Filters, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Accessories for Gas Clean Purification System, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® ETFE Fluoropolymer Tubing, IDEX Health & Science
: Upchurch Scientific
pH Gradient Buffers
: Thermo Scientific
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Agilent Autosampler Supplies, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Graphite Ferrules for GC
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)