$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Febuxostat n-Butyl Isomer
TRC Febuxostat n-Butyl Isomer
77984-619
:
LGC Standards
New Product
:
TRC Febuxostat n-Butyl Isomer
77984-619
:
LGC Standards
:
TRC-F229035-100MG
New Product
:
CAS No.:
1657014-33-9
Description:
Febuxostat n-Butyl Isomer
Size:
100 mg