$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Noscapine Hydrochloride
TRC Noscapine Hydrochloride
TRC Noscapine Hydrochloride
Catalog # 77990-674
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Noscapine Hydrochloride
Catalog # 77990-674
Supplier:
LGC Standards
Supplier Number:
TRC-N882018-100MG
New Product
CAS Number:
Specifications
CAS No.:
912-60-7
Description:
Noscapine Hydrochloride
Size:
100 mg
Specifications
About this item
Product Details
Product Details