Order Entry
Puerto Rico
ContactUsLinkComponent
CHEMets® Chlorine Visual HR Test 0 to 25 ppm
CHEMets® Chlorine Visual HR Test 0 to 25 ppm
Catalog # 77674-760
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
CHEMets® Chlorine Visual HR Test 0 to 25 ppm
Catalog # 77674-760
Supplier:  CHEMetrics
Supplier Number:  K-2520D
New Product
CAS Number:  

Specifications

  • Chemical method:
    DPD (free and total)
  • Application:
    Drinking water;Waste water;Surface and ground water

Specifications

About this item

Delivery: The K-2520D chlorine HR CHEMets® test kit (0 to 25 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): R to 2500 CHEMets®refill, C to 2506 Comparato r, A to 2500 activato r solution, 25 ml sample cup, A to 0104 10ml syringe, and instructions.