Filming Amines Instrumental Test Kit
Catalog # 77674-714
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Rose Bengal
- Cat. No.:77674-714
- Application:Boiler water
- Range:0 - 6.00 ppm as ODA
Specifications
About this item
Film-forming amines, also known as FFAP or FFP, are chemicals applied in high-pressure boilers, cooling systems, and HRSGs to guard against corrosion and reduce the presence of corrosive agents. The K-1013 test kit utilizes the Rose Bengal method to detect and measure these substances in boiler water or steam condensate.
- Range: 0 to 6.00 ppm as ODA Method: Rose Bengal Analysis time: 2 minutes
This kit requires the use of a V-3000 multi-analyte photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell.
Delivery: K-1013 kit comes in a cardboard box and contains everything needed to perform 30 tests: thirty ampoules, acidifier solution, 25 ml sample cup, ampoule blank, and instructions.