Nitrite Acidifier Solution Pack
77674-576
New Product
:
- Chemical method:Azo Dye Formation
The A-7004 accessory pack supplies enough Nitrite Acidifier Solution to perform approximately 200 tests. Quantity per pack: Six 10 ml Bottles.
The A-7004 accessory pack supplies enough Nitrite Acidifier Solution to perform approximately 200 tests. Quantity per pack: Six 10 ml Bottles.