Paragon Scientific Type PSU0600, Viscosity Reference Standard
77975-919
New Product
:
- Description:Type PSU0600, viscosity reference standard
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
- Standard type: Single component