Order Entry
Puerto Rico
ContactUsLinkComponent
Lipomed PCP-D5.HCl (Phencyclidine-D5.HCl), 1 mg/ml in Methanol (as free base)
Lipomed PCP-D5.HCl (Phencyclidine-D5.HCl), 1 mg/ml in Methanol (as free base)
  77976-229
New Product
 :  
undefined
Lipomed PCP-D5.HCl (Phencyclidine-D5.HCl), 1 mg/ml in Methanol (as free base)
  77976-229
 :  LPM-PCP-437-HC-1LM
New Product
 :  

 

  • Matrix:
    Methanol
  • CAS No.:
    60124-79-0
  • Description:
    PCP-D5.HCl (Phencyclidine-D5.HCl), 1 mg/ml in Methanol (as free base)
  • Size:
    1 ml