Order Entry
Puerto Rico
ContactUsLinkComponent
TRC 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
TRC 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
Catalog # 77994-096
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
TRC 1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
Catalog # 77994-096
Supplier:  LGC Standards
Supplier Number:  TRC-T293930-1G
New Product
CAS Number:  

Specifications

  • CAS No.:
    99614-64-9
  • Description:
    1,2,3,9-Tetrahydro-9-methyl-3-methylene-4H-carbazol-4-one
  • Size:
    1 g

Specifications

About this item