$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC p-Toluic Acid
TRC p-Toluic Acid
77994-473
:
LGC Standards
New Product
:
TRC p-Toluic Acid
77994-473
:
LGC Standards
:
TRC-T536125-5G
New Product
:
CAS No.:
99-94-5
Description:
p-Toluic Acid
Size:
5 g