Paragon Scientific Sulfur @ 0.0005% (5 ppm) in light mineral oil
77975-868
New Product
:
- Matrix:Light Mineral Oil
- CAS No.:7704-34-9
- Description:Sulfur @ 0.0005% (5 ppm) in light mineral oil
- Size:100 ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065