Paragon Scientific ASTM D7220, Sulfur set B in synthetic diesel: S @ 0.005, 0.01, 0.025, 0.05, 0.075, 0.1%
Catalog # 77975-621
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Synthetic Diesel
- CAS No.:7704-34-9
- Description:ASTM D7220, Sulfur set B in synthetic diesel: S @ 0.005, 0.01, 0.025, 0.05, 0.075, 0.1%
- Size:6×100ml
- Certification:ISO 17034
- Storage Conditions:RT
- Shipping Conditions:RT
- Molecular Weight:32.065